Assay Details
| Gene Symbol: Ctsc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017097.1 | NP_058793.1 | 4 - 5 | 702 | 91 | |
| GenBank mRNA | D90404.1 | - | 4 - 5 | 702 | 91 | |
| FQ218311.1 | - | 762 | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
T cell mediated cytotoxicity
proteolysis
apoptotic process
aging
response to organic substance
positive regulation of peptidase activity
positive regulation of proteolysis involved in cellular protein catabolic process
positive regulation of apoptotic signaling pathway
cysteine-type endopeptidase activity
serine-type endopeptidase activity
cysteine-type peptidase activity
peptidase activator activity involved in apoptotic process
phosphatase binding
chloride ion binding
identical protein binding
protein self-association
chaperone binding
T cell mediated cytotoxicity
proteolysis
apoptotic process
aging
response to organic substance
positive regulation of peptidase activity
positive regulation of proteolysis involved in cellular protein catabolic process
positive regulation of apoptotic signaling pathway
cysteine-type endopeptidase activity
serine-type endopeptidase activity
cysteine-type peptidase activity
peptidase activator activity involved in apoptotic process
phosphatase binding
chloride ion binding
identical protein binding
protein self-association
chaperone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P11819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344140 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132350 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127368 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576548 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748207 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235795 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106200 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378450 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146686 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12543 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72459 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11819 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P11819 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P11819 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11819 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76800 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33799 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 3078 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10708523 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708530 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708529 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708526 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708527 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708522 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708524 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708528 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10708521 | - | - |
| Rat Genome 230 2.0 Array | 1374778_at | - | - |
| Rat Genome 230 2.0 Array | 1368280_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1374778_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368280_at | - | - |
| Affymetrix HT Rat Focus Array | 1368280_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368280_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100015390.rn.1 | TC0100001975.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100015388.rn.1 | TC0100001975.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100015389.rn.1 | TC0100001975.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100015391.rn.1 | TC0100001975.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0100015385.rn.1 | TC0100001975.rn.1 | RTA TC |
| Rat Expression Set 230 | 1374778_PM_at | - | - |
| Rat Expression Set 230 | 1374778_at | - | - |
| Rat Expression Set 230 | 1368280_at | - | - |
| Rat Expression Set 230 | 1368280_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10708521 | - | - |
| Rat Genome U34 Set | D90404_g_at | - | - |
| Rat Genome U34 Set | D90404_at | - | - |
| Rat Genome U34 Set | rc_AA926360_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21660673 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359951 | - | - |
| RatRef-12 Expression BeadChip | ILMN_62796 | - | - |

Ctsc
Genomic Map