Assay Details
| Gene Symbol: Pla2g10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017176.2 | NP_058872.1 | 1 - 2 | 101 | 107 | |
| GenBank mRNA | AF166100.1 | - | 1 - 2 | 103 | 107 | |
| AY651029.1 | - | 1 - 2 | 97 | 107 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF166100.1, AY651029.1, NM_017176.2 transcripts.
Set Membership: |
|

Pla2g10
Genomic Map