Assay Details
| Gene Symbol: Cxcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017183.1 | NP_058879.1 | 1 - 2 | 114 | 66 | |
| GenBank mRNA | X77797.1 | - | 1 - 2 | 114 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_017183.1, X77797.1 transcripts.
Set Membership: |
|

Cxcr2
Genomic Map