Assay Details
| Gene Symbol: Aif1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017196.3 | NP_058892.1 | 4 - 5 | 289 | 86 | |
| XM_006256065.2 | XP_006256127.1 | 6 - 7 | 439 | 86 | ||
| GenBank mRNA | AB000818.1 | - | 4 - 5 | 287 | 86 | |
| D82069.1 | - | 6 - 7 | 444 | 86 | ||
| FM053942.1 | - | 4 - 5 | 276 | 86 | ||
| FQ231499.1 | - | 4 - 5 | 287 | 86 | ||
| U17919.1 | - | 4 - 5 | 261 | 86 | ||
More Information
Set Membership: |
|

Aif1
Genomic Map