Assay Details
| Gene Symbol: Pde3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017229.1 | NP_058925.1 | 12 - 13 | 2534 | 83 | |
| GenBank mRNA | Z22867.1 | - | 12 - 13 | 2534 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
cAMP catabolic process
negative regulation of cell adhesion
signal transduction
diterpenoid metabolic process
negative regulation of angiogenesis
endocrine pancreas development
cellular response to insulin stimulus
glucose homeostasis
negative regulation of insulin secretion
regulation of insulin secretion
negative regulation of lipid catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
drug binding
cAMP binding
protein kinase B binding
metal ion binding
phosphoprotein binding
angiogenesis
cAMP catabolic process
negative regulation of cell adhesion
signal transduction
diterpenoid metabolic process
negative regulation of angiogenesis
endocrine pancreas development
cellular response to insulin stimulus
glucose homeostasis
negative regulation of insulin secretion
regulation of insulin secretion
negative regulation of lipid catabolic process
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
cGMP-inhibited cyclic-nucleotide phosphodiesterase activity
drug binding
cAMP binding
protein kinase B binding
metal ion binding
phosphoprotein binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1004645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306933 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277641 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365213 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 774424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125583 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236027 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416975 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30029 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726436 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506236 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526448 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69175 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689850 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 893993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1130 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670724 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916826 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573625 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491167 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410969 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761978 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474666 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780963 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552811 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191571 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 209825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448084 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95590 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439866 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60382 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520780 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330097 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641232 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545549 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344174 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170870 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730922 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352723 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16505 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59112 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157014 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141024 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131008 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155407 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1004645 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1004645 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1004645 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1004645 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88274 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19212 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 11324 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 27430 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102570 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 87799 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 63021 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104704 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10710093 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710096 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710097 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710098 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710104 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710095 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710092 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710105 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710100 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710103 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710102 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710101 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710090 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710091 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710099 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710094 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10710089 | - | - |
| Rat Genome 230 2.0 Array | 1369157_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369157_at | - | - |
| Affymetrix HT Rat Focus Array | 1369157_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369157_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100018412.rn.1 | TC0100002430.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369157_at | - | - |
| Rat Expression Set 230 | 1369157_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10710089 | - | - |
| Rat Genome U34 Set | Z22867_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22262565 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363159 | - | - |
| RatRef-12 Expression BeadChip | ILMN_51892 | - | - |

Pde3b
Genomic Map