Assay Details
| Gene Symbol: Gclm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017305.2 | NP_059001.1 | 1 - 2 | 251 | 80 | |
| GenBank mRNA | BC078867.1 | - | 1 - 2 | 251 | 7110139 | 80 |
| L22191.1 | - | 1 - 2 | 189 | 80 | ||
| S65555.1 | - | 1 - 2 | 189 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gclm
Genomic Map