Assay Details
| Gene Symbol: Gnas | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024823.4 | NP_001019994.4 | 2 - 3 | 2466 | 84 | |
| NM_001159653.1 | NP_001153125.1 | 1135 | 84 | |||
| NM_001159656.1 | NP_001153128.1 | 3 - 4 | 1135 | 84 | ||
| NM_019132.1 | NP_062005.1 | 395 | 84 | |||
| NM_021845.5 | NP_068617.6 | 2 - 3 | 2466 | 84 | ||
| GenBank mRNA | AF105254.1 | - | 993 | 84 | ||
| AF107844.1 | - | 953 | 84 | |||
| AF107845.1 | - | 3 - 4 | 1094 | 84 | ||
| BC061967.1 | - | 357 | 84 | |||
| BC158589.1 | - | 250 | 9099119 | 84 | ||
| BC168156.1 | - | 259 | 84 | |||
| DQ120475.1 | - | 244 | 84 | |||
| DQ120476.1 | - | 244 | 84 | |||
| FQ214546.1 | - | 134 | 84 | |||
| L10326.1 | - | 2 - 3 | 231 | 84 | ||
| M12673.1 | - | 459 | 84 | |||
| M17525.1 | - | 395 | 84 | |||
| X84047.1 | - | 2 - 3 | 1614 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnas
Genomic Map