Assay Details
| Gene Symbol: Gdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019139.1 | NP_062012.1 | 2 - 3 | 204 | 122 | |
| GenBank mRNA | AF205713.1 | - | 2 - 3 | 208 | 122 | |
| AF497634.1 | - | 1 - 2 | 132 | 122 | ||
| EU068469.1 | - | 1 - 2 | 170 | 122 | ||
| EU068470.1 | - | 1 - 2 | 155 | 122 | ||
| EU068471.1 | - | 1 - 2 | 155 | 122 | ||
| EU068472.1 | - | 1 - 2 | 155 | 122 | ||
| L15305.1 | - | 2 - 3 | 204 | 122 | ||
| S75583.1 | - | 1 - 2 | 155 | 122 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gdnf
Genomic Map