Assay Details
Gene Symbol: Cyp2c11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019184.2 | NP_062057.2 | 5 - 6 | 836 | 73 | |
XM_008760364.1 | XP_008758586.1 | 4 - 5 | 664 | 73 | ||
GenBank mRNA | BC088146.1 | - | 5 - 6 | 826 | 7369647 | 73 |
J02657.1 | - | 5 - 6 | 836 | 73 | ||
U33173.1 | - | 5 - 6 | 836 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
