Assay Details
| Gene Symbol: Neurog1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019207.1 | NP_062080.1 | 1 | 353 | 73 | |
| GenBank mRNA | U67777.1 | - | 1 | 353 | 73 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Neurog1
Genomic Map