Assay Details
Gene Symbol: Dync1h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019226.3 | NP_062099.3 | 11 - 12 | 3127 | 60 | |
GenBank mRNA | L08505.1 | - | 11 - 12 | 3127 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P151408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 336354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 531383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607031 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31068 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 37376 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151408 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P151408 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P151408 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P348868 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P552706 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20555 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30203 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 81256 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58082 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63826 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90222 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10887147 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887167 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887177 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887179 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887159 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887161 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887207 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887155 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887169 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887165 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887191 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887194 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887163 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887178 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887182 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887195 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887199 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887156 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887212 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887187 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887218 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887162 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887176 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887205 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887198 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887219 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887193 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887173 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887158 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887154 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887210 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887208 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887196 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887213 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887186 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887200 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887180 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887215 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887168 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887202 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887146 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887175 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887174 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887183 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887203 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887166 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887192 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887214 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887172 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887157 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887181 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887217 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887145 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887160 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887211 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887206 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887171 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887190 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887189 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887197 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887184 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887188 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887185 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887209 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887201 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887164 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887170 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887204 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887220 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10887144 | - | - |
Rat Genome 230 2.0 Array | 1398803_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398803_at | - | - |
Affymetrix HT Rat Focus Array | 1398803_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398803_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600010784.rn.1 | TC0600001563.rn.1 | RTA TC |
Rat Expression Set 230 | 1398803_at | - | - |
Rat Expression Set 230 | 1398803_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10887144 | - | - |
Rat Genome U34 Set | L08505_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21443588 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376210 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376672 | - | - |
