Assay Details
Gene Symbol: Itsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136096.1 | NP_001129568.1 | 245 | 60 | ||
NM_019227.1 | NP_062100.1 | 3 - 4 | 224 | 60 | ||
GenBank mRNA | AF127798.1 | - | 3 - 4 | 224 | 60 | |
AF132672.1 | - | 3 - 4 | 224 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
endocytosis
small GTPase mediated signal transduction
positive regulation of phosphorylation
positive regulation of catalytic activity
negative regulation of neuron apoptotic process
positive regulation of protein kinase B signaling
calcium ion binding
protein binding
kinase activator activity
protein complex scaffold
proline-rich region binding
exocytosis
endocytosis
small GTPase mediated signal transduction
positive regulation of phosphorylation
positive regulation of catalytic activity
negative regulation of neuron apoptotic process
positive regulation of protein kinase B signaling
calcium ion binding
protein binding
kinase activator activity
protein complex scaffold
proline-rich region binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P034384 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P138115 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P247981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530547 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 20722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582704 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 206379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 166934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136098 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 937725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694117 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699258 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903190 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 892058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104827 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127302 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173617 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034384 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P138115 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P247981 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P247981 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P034384 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P138115 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P247981 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P138115 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P034384 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P247981 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P11984 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701746334 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11874 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71995 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46293 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701746334 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10750235 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750246 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750268 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750232 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750258 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750263 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750237 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750259 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750234 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750267 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750273 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750272 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750238 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750269 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750233 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750248 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750275 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750243 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750271 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750236 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750239 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750247 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750257 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750266 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750245 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750262 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750256 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750241 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750270 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750250 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750231 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750244 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750260 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750265 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750251 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750261 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750264 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750242 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750276 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750249 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750240 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750274 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750253 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750252 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10750230 | - | - |
Rat Genome 230 2.0 Array | 1397161_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1378817_at | - | - |
Rat Genome 230 2.0 Array | 1395008_at | - | - |
Rat Genome 230 2.0 Array | 1397161_a_at | - | - |
Rat Genome 230 2.0 Array | 1395008_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378817_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378817_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1378817_at | - | - |
Affymetrix HT Rat Focus Array | 1395008_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1397161_a_at | - | - |
Affymetrix HT Rat Focus Array | 1397161_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1395008_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100001086.rn.1 | TC1100000258.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100001087.rn.1 | TC1100000258.rn.1 | RTA TC |
Rat Expression Set 230 | 1397161_a_at | - | - |
Rat Expression Set 230 | 1397161_PM_a_at | - | - |
Rat Expression Set 230 | 1395008_PM_at | - | - |
Rat Expression Set 230 | 1378817_PM_at | - | - |
Rat Expression Set 230 | 1395008_at | - | - |
Rat Expression Set 230 | 1378817_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10750230 | - | - |
Rat Genome U34 Set | rc_AI071501_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21623784 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352431 | - | - |
