Assay Details
Gene Symbol: Sgk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193568.1 | NP_001180497.1 | 8 - 9 | 936 | 79 | |
NM_001193569.1 | NP_001180498.1 | 8 - 9 | 874 | 79 | ||
NM_019232.3 | NP_062105.2 | 8 - 9 | 848 | 79 | ||
XM_006227723.1 | XP_006227785.1 | 10 - 11 | 1534 | 79 | ||
XM_008758606.1 | XP_008756828.1 | 8 - 9 | 1044 | 79 | ||
GenBank mRNA | BC078843.1 | - | 8 - 9 | 802 | 7130800 | 79 |
L01624.1 | - | 8 - 9 | 853 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
