Assay Details
Gene Symbol: Ifrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019242.1 | NP_062115.1 | 9 - 10 | 1180 | 84 | |
XM_008764643.1 | XP_008762865.1 | 8 - 9 | 1799 | 84 | ||
GenBank mRNA | BC083560.1 | - | 9 - 10 | 1246 | 7193329 | 84 |
J04511.1 | - | 9 - 10 | 1180 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
