Assay Details
Gene Symbol: Slc22a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019269.1 | NP_062142.1 | 4 - 5 | 928 | 95 | |
GenBank mRNA | AF110416.1 | - | 4 - 5 | 928 | 95 | |
AJ001933.1 | - | 4 - 5 | 914 | 95 | ||
BC166421.1 | - | 3 - 4 | 677 | 9089728 | 95 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
