Assay Details
| Gene Symbol: Slc14a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019347.2 | NP_062220.2 | 7 - 8 | 1050 | 73 | |
| NM_177962.3 | NP_808877.2 | 7 - 8 | 1050 | 73 | ||
| GenBank mRNA | AF031642.1 | - | 7 - 8 | 1050 | 73 | |
| AF041788.1 | - | 7 - 8 | 1050 | 73 | ||
| AF042167.1 | - | 7 - 8 | 1050 | 73 | ||
| U77971.1 | - | 7 - 8 | 1050 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc14a2
Genomic Map