Assay Details
| Gene Symbol: Syt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019350.2 | NP_062223.1 | 2 - 3 | 401 | 68 | |
| XM_006228287.2 | XP_006228349.1 | 1 - 2 | 91 | 68 | ||
| GenBank mRNA | BC092198.1 | - | 2 - 3 | 401 | 68 | |
| U26402.1 | - | 2 - 3 | 204 | 68 | ||
| X84884.1 | - | 1 - 2 | 154 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Syt5
Genomic Map