Assay Details
| Gene Symbol: Enpp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019370.2 | NP_062243.2 | 20 - 21 | 2008 | 67 | |
| GenBank mRNA | BC097326.1 | - | 20 - 21 | 2008 | 7458431 | 67 |
| D30649.1 | - | 20 - 21 | 67 | |||
| FQ218696.1 | - | 3 - 4 | 393 | 67 | ||
| U78787.1 | - | 20 - 21 | 67 | |||
| U78788.1 | - | 20 - 21 | 2011 | 67 | ||
| Z47987.1 | - | 20 - 21 | 1992 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
phosphate-containing compound metabolic process
receptor-mediated endocytosis
immune response
nucleoside triphosphate catabolic process
inorganic diphosphate transport
regulation of smooth muscle cell differentiation
nucleic acid phosphodiester bond hydrolysis
nucleic acid binding
phosphodiesterase I activity
nucleotide diphosphatase activity
scavenger receptor activity
polysaccharide binding
NADH pyrophosphatase activity
metal ion binding
nucleoside-triphosphate diphosphatase activity
phosphate-containing compound metabolic process
receptor-mediated endocytosis
immune response
nucleoside triphosphate catabolic process
inorganic diphosphate transport
regulation of smooth muscle cell differentiation
nucleic acid phosphodiester bond hydrolysis
nucleic acid binding
phosphodiesterase I activity
nucleotide diphosphatase activity
scavenger receptor activity
polysaccharide binding
NADH pyrophosphatase activity
metal ion binding
nucleoside-triphosphate diphosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P698409 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238585 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820218 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441212 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116392 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755531 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839012 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306374 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875697 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944016 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 61502 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916766 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846582 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230765 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239336 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19102 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280901 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547275 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620211 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917317 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962172 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935773 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102561 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409309 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62161 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98856 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144970 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P698409 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P698409 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P698409 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P501103 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P698409 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701484394 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701880513 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 82855 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 14571 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95790 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 104705 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7626 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701484394 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701880513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10702238 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702241 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702245 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702249 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702231 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702232 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702242 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702229 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702236 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702243 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702246 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702226 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702225 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702235 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702230 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702248 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702233 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702247 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702227 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702228 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702234 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702239 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702244 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702240 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702237 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10702224 | - | - |
| Rat Genome 230 2.0 Array | 1367905_at | - | - |
| Rat Genome 230 2.0 Array | 1367905_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367905_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367905_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100001312.rn.1 | TC0100000231.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367905_at | - | - |
| Rat Expression Set 230 | 1367905_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10702224 | - | - |
| Rat Genome U34 Set | D30649mRNA_s_at | - | - |
| Rat Genome U34 Set | rc_AI178131_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21938067 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530484 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376301 | - | - |
| RatRef-12 Expression BeadChip | ILMN_160691 | - | - |

Enpp3
Genomic Map