Assay Details
| Gene Symbol: Srcin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019378.1 | NP_062251.1 | 3496 | 85 | ||
| GenBank mRNA | AF156981.1 | - | 3496 | 85 | ||
| AF156982.1 | - | 3496 | 85 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
negative regulation of cell adhesion
regulation of cell migration
substrate adhesion-dependent cell spreading
negative regulation of protein secretion
regulation of dendritic spine morphogenesis
positive regulation of protein tyrosine kinase activity
negative regulation of protein tyrosine kinase activity
protein binding
protein kinase binding
protein domain specific binding
exocytosis
negative regulation of cell adhesion
regulation of cell migration
substrate adhesion-dependent cell spreading
negative regulation of protein secretion
regulation of dendritic spine morphogenesis
positive regulation of protein tyrosine kinase activity
negative regulation of protein tyrosine kinase activity
protein binding
protein kinase binding
protein domain specific binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P109984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179767 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141245 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721872 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668310 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463473 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546862 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784831 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874099 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 513993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158627 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345250 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669863 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414665 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663139 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403804 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813649 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541216 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39063 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140080 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75101 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109984 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P109984 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P109984 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P416871 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701737452 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 102123 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 72313 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 35213 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 12413 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 17343 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701737452 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10746742 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746731 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746738 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746729 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746737 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746730 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746734 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746736 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746726 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746723 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746739 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746727 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746724 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746725 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746741 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746732 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746735 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746733 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10746721 | - | - |
| Rat Genome 230 2.0 Array | 1370655_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1369340_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369340_at | - | - |
| Rat Genome 230 2.0 Array | 1370655_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1370655_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1369340_at | - | - |
| Affymetrix HT Rat Focus Array | 1370655_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1369340_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000032885.rn.1 | TC1000003686.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1000032886.rn.1 | TC1000003686.rn.1 | RTA TC |
| Rat Expression Set 230 | 1370655_a_at | - | - |
| Rat Expression Set 230 | 1369340_PM_at | - | - |
| Rat Expression Set 230 | 1369340_at | - | - |
| Rat Expression Set 230 | 1370655_PM_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10746721 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21409708 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1361659 | - | - |
| RatRef-12 Expression BeadChip | ILMN_49321 | - | - |

Srcin1
Genomic Map