Assay Details
Gene Symbol: Parva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020656.2 | NP_065707.2 | 12 - 13 | 1039 | 66 | |
GenBank mRNA | AF264765.2 | - | 12 - 13 | 1039 | 66 | |
FQ228521.1 | - | 1099 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sprouting angiogenesis
outflow tract septum morphogenesis
cell adhesion
establishment or maintenance of cell polarity
regulation of cell shape
actin cytoskeleton organization
actin cytoskeleton reorganization
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
protein heterooligomerization
cilium morphogenesis
actin-mediated cell contraction
smooth muscle cell chemotaxis
actin binding
protein domain specific binding
sprouting angiogenesis
outflow tract septum morphogenesis
cell adhesion
establishment or maintenance of cell polarity
regulation of cell shape
actin cytoskeleton organization
actin cytoskeleton reorganization
heterotypic cell-cell adhesion
substrate adhesion-dependent cell spreading
protein heterooligomerization
cilium morphogenesis
actin-mediated cell contraction
smooth muscle cell chemotaxis
actin binding
protein domain specific binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P026509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 551809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 146519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804682 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178355 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491298 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141533 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741916 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 960753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 500065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 92434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165852 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 2449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139901 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026509 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P026509 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P026509 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15542 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 79083 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 69340 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14501 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37694 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3103 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15826 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 86161 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65223 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 22223 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92760 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10710002 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710003 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10709996 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710000 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10709999 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710001 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710004 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10709998 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10709997 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10709995 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10709994 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10710005 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10709993 | - | - |
Rat Genome 230 2.0 Array | 1370266_at | - | - |
Rat Genome 230 2.0 Array | 1370266_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371588_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371588_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100018169.rn.1 | TC0100002385.rn.1 | RTA TC |
Rat Expression Set 230 | 1370266_at | - | - |
Rat Expression Set 230 | 1370266_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10709993 | - | - |
Rat Genome U34 Set | rc_AA998633_at | - | - |
Rat Genome U34 Set | rc_AI237885_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21598674 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369439 | - | - |
RatRef-12 Expression BeadChip | ILMN_50655 | - | - |
