Assay Details
| Gene Symbol: Ltbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021586.2 | NP_067597.2 | 18 - 19 | 2991 | 57 | |
| XM_006240350.2 | XP_006240412.1 | 18 - 19 | 3183 | 57 | ||
| GenBank mRNA | Y12760.1 | - | 18 - 19 | 2990 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ltbp2
Genomic Map