Assay Details
| Gene Symbol: Ltb4r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021656.1 | NP_067688.1 | 1 | 751 | 85 | |
| XM_006252011.2 | XP_006252073.1 | 2 | 2425 | 85 | ||
| XM_006252012.1 | XP_006252074.1 | 2 | 787 | 85 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ltb4r
Genomic Map