Assay Details
| Gene Symbol: Jun | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021835.3 | NP_068607.1 | 1 | 1254 | 53 | |
| GenBank mRNA | BC078738.1 | - | 1 | 491 | 7124370 | 53 |
| X17163.1 | - | 1 | 748 | 53 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Jun
Genomic Map