Assay Details
| Gene Symbol: Kit | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022264.1 | NP_071600.1 | 1 - 2 | 116 | 68 | |
| XM_006250909.2 | XP_006250971.1 | 1 - 2 | 150 | 68 | ||
| GenBank mRNA | AF228308.1 | - | 1 - 2 | 93 | 68 | |
| AF228310.1 | - | 1 - 2 | 93 | 68 | ||
| AF228311.1 | - | 1 - 2 | 93 | 68 | ||
| AF296694.1 | - | 1 - 2 | 100 | 68 | ||
| D12524.1 | - | 1 - 2 | 116 | 68 | ||
| EU247827.1 | - | 1 - 2 | 100 | 68 | ||
| EU247828.1 | - | 1 - 2 | 100 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kit
Genomic Map