Assay Details
| Gene Symbol: Mre11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022279.1 | NP_071615.1 | 13 - 14 | 1576 | 75 | |
| XM_006242532.2 | XP_006242594.1 | 13 - 14 | 1671 | 75 | ||
| XM_006242533.1 | XP_006242595.1 | 13 - 14 | 1665 | 75 | ||
| XM_006242534.2 | XP_006242596.1 | 14 - 15 | 1747 | 75 | ||
| GenBank mRNA | AF218574.1 | - | 13 - 14 | 1576 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
telomere maintenance
base-excision repair
nucleotide-excision repair
double-strand break repair
cellular response to DNA damage stimulus
sister chromatid cohesion
mitotic G2 DNA damage checkpoint
synapsis
heart development
cell proliferation
intra-S DNA damage checkpoint
positive regulation of protein autophosphorylation
DNA duplex unwinding
negative regulation of DNA endoreduplication
positive regulation of kinase activity
negative regulation of viral entry into host cell
chromosome organization
meiotic cell cycle
nucleic acid phosphodiester bond hydrolysis
DNA binding
ATP-dependent DNA helicase activity
endonuclease activity
exonuclease activity
protein binding
protein C-terminus binding
3'-5' exonuclease activity
manganese ion binding
telomere maintenance
base-excision repair
nucleotide-excision repair
double-strand break repair
cellular response to DNA damage stimulus
sister chromatid cohesion
mitotic G2 DNA damage checkpoint
synapsis
heart development
cell proliferation
intra-S DNA damage checkpoint
positive regulation of protein autophosphorylation
DNA duplex unwinding
negative regulation of DNA endoreduplication
positive regulation of kinase activity
negative regulation of viral entry into host cell
chromosome organization
meiotic cell cycle
nucleic acid phosphodiester bond hydrolysis
DNA binding
ATP-dependent DNA helicase activity
endonuclease activity
exonuclease activity
protein binding
protein C-terminus binding
3'-5' exonuclease activity
manganese ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P436859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954274 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368364 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709458 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139424 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929002 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698711 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624298 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187220 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880951 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703659 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461979 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304774 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 722106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815463 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120203 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138600 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136264 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132305 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81870 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P436859 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P436859 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P436859 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P436859 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 7986 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 2893 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76741 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 33071 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10908056 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908059 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908063 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908065 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908062 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908064 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908061 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908057 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908060 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908071 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908070 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908067 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908054 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908066 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908069 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908068 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908055 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10908053 | - | - |
| Rat Genome 230 2.0 Array | 1369445_at | - | - |
| Rat Genome 230 2.0 Array | 1369445_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369445_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369445_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0800000670.rn.1 | TC0800000110.rn.1 | RTA TC |
| Rat Expression Set 230 | 1369445_PM_at | - | - |
| Rat Expression Set 230 | 1369445_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10908053 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21281957 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373654 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69827 | - | - |

Mre11a
Genomic Map