Assay Details
| Gene Symbol: Clec10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022393.2 | NP_071788.2 | 1 - 2 | 185 | 59 | |
| XM_006246803.2 | XP_006246865.1 | 1 - 2 | 220 | 59 | ||
| XM_008767867.1 | XP_008766089.1 | 1 - 2 | 220 | 59 | ||
| XM_008767868.1 | XP_008766090.1 | 1 - 2 | 64 | 59 | ||
| XM_008767869.1 | XP_008766091.1 | 1 - 2 | 64 | 59 | ||
| GenBank mRNA | BC091151.1 | - | 1 - 2 | 185 | 7375234 | 59 |
| FQ213042.1 | - | 1 - 2 | 54 | 59 | ||
| FQ228437.1 | - | 1 - 2 | 43 | 59 | ||
| J05495.1 | - | 1 - 2 | 45 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC091151.1, J05495.1, NM_022393.2, XM_006246803.2, XM_008767867.1, XM_008767868.1, XM_008767869.1 transcripts.
Set Membership: |
|

Clec10a
Genomic Map