Assay Details
Gene Symbol: Phka1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022626.1 | NP_072148.1 | 30 - 31 | 3302 | 121 | |
XM_006257101.2 | XP_006257163.1 | 29 - 30 | 3590 | 121 | ||
XM_006257102.2 | XP_006257164.1 | 29 - 30 | 3452 | 121 | ||
GenBank mRNA | AF197561.1 | - | 30 - 31 | 3302 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P181039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414061 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 208111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499478 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 462057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 35662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494156 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 835656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326354 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 800515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57332 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 602116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189544 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503410 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19212 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 44328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 55421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26900 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P181039 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P181039 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P181039 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P181039 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29208 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43443 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 32925 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 31851 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88160 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61261 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64597 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102240 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104333 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 51998 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10938760 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938767 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938764 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938762 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938769 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938753 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938759 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938757 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938771 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938749 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938770 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938761 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938750 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938751 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938755 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938768 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938756 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938765 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938748 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938754 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938766 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938772 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938758 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938752 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938763 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10938747 | - | - |
Rat Genome 230 2.0 Array | 1369357_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390210_PM_at | - | - |
Rat Genome 230 2.0 Array | 1390210_at | - | - |
Rat Genome 230 2.0 Array | 1369357_at | - | - |
Affymetrix HT Rat Focus Array | 1390210_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369357_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1390210_at | - | - |
Affymetrix HT Rat Focus Array | 1369357_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00012501.rn.1 | TC0X00001964.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00012499.rn.1 | TC0X00001964.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00012500.rn.1 | TC0X00001964.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00012498.rn.1 | TC0X00001964.rn.1 | RTA TC |
Rat Expression Set 230 | 1390210_PM_at | - | - |
Rat Expression Set 230 | 1369357_at | - | - |
Rat Expression Set 230 | 1390210_at | - | - |
Rat Expression Set 230 | 1369357_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10938747 | - | - |
Rat Genome U34 Set | M92919_at | - | - |
Rat Genome U34 Set | rc_AA817929_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20998431 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365657 | - | - |
RatRef-12 Expression BeadChip | ILMN_57999 | - | - |
