Assay Details
| Gene Symbol: Tmprss11d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033652.1 | NP_001028824.1 | 9 - 10 | 1342 | 80 | |
| NM_022630.1 | NP_072152.1 | 4 - 5 | 713 | 80 | ||
| GenBank mRNA | AF198087.1 | - | 4 - 5 | 713 | 80 | |
| AF453776.1 | - | 9 - 10 | 1342 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P032323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111051 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80657 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432423 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 109934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972495 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312166 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324111 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630090 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143701 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80246 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55890 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759425 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 449289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386160 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222971 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864516 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168254 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113468 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73735 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51503 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P032323 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P032323 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P032323 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P188047 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34346 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 9188 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76304 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 81903 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 75309 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 85280 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46587 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10772046 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772047 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772049 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772052 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772043 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772048 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772044 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772045 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772051 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772054 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772053 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772050 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10772042 | - | - |
| Rat Genome 230 2.0 Array | 1387620_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1387620_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1387620_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1387620_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1400002675.rn.1 | TC1400000325.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1400002676.rn.1 | TC1400000325.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387620_PM_a_at | - | - |
| Rat Expression Set 230 | 1387620_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10772042 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 20890797 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371717 | - | - |
| RatRef-12 Expression BeadChip | ILMN_59775 | - | - |

Tmprss11d
Genomic Map