Assay Details
| Gene Symbol: Bid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022684.1 | NP_073175.1 | 1 - 2 | 96 | 80 | |
| GenBank mRNA | AF259503.1 | - | 1 - 2 | 96 | 80 | |
| D89804.1 | - | 52 | 80 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF259503.1, D89804.1, NM_022684.1 transcripts.
Set Membership: |
|

Bid
Genomic Map