Assay Details
| Gene Symbol: Nab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006244902.2 | XP_006244964.1 | 2 - 3 | 1094 | 85 | |
| GenBank mRNA | FQ222344.1 | - | 3 - 4 | 1070 | 85 | |
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P470808 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654219 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275604 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438114 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147587 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 627567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 370155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536995 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692817 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578201 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101030 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 11284 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211897 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582145 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145481 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79253 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61194 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10924 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76324 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P470808 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P470808 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P470808 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P470808 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44055 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 44319 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66981 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 22834 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10923204 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923199 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923203 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923202 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923200 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923201 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923206 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923207 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923205 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10923198 | - | - |
| Rat Genome 230 2.0 Array | 1393172_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1369409_at | - | - |
| Rat Genome 230 2.0 Array | 1369409_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1393172_at | - | - |
| Affymetrix HT Rat Focus Array | 1369409_at | - | - |
| Affymetrix HT Rat Focus Array | 1393172_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1369409_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1393172_at | - | - |
| Rat Expression Set 230 | 1369409_at | - | - |
| Rat Expression Set 230 | 1369409_PM_at | - | - |
| Rat Expression Set 230 | 1393172_at | - | - |
| Rat Expression Set 230 | 1393172_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10923198 | - | - |
| Rat Genome U34 Set | U17253_at | - | - |
| Rat Genome U34 Set | rc_AA848646_at | - | - |
| Rat Genome U34 Set | rc_AI236912_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22161648 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1352667 | - | - |
| RatRef-12 Expression BeadChip | ILMN_69831 | - | - |

Nab1
Genomic Map