Assay Details
Gene Symbol: Slc13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022866.2 | NP_074057.2 | 12 - 13 | 1653 | 70 | |
GenBank mRNA | AF080451.1 | - | 12 - 13 | 1630 | 70 | |
AF081825.1 | - | 12 - 13 | 1655 | 70 | ||
BC129094.1 | - | 12 - 13 | 1653 | 8365494 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sodium ion transport
dicarboxylic acid transport
succinate transport
citrate transport
aspartate transport
sodium ion transmembrane transport
transmembrane transport
succinate transmembrane transport
L-aspartate transmembrane transport
dicarboxylic acid transmembrane transporter activity
organic acid:sodium symporter activity
citrate transmembrane transporter activity
succinate transmembrane transporter activity
L-aspartate transmembrane transporter activity
high-affinity sodium:dicarboxylate symporter activity
sodium:dicarboxylate symporter activity
sodium ion transport
dicarboxylic acid transport
succinate transport
citrate transport
aspartate transport
sodium ion transmembrane transport
transmembrane transport
succinate transmembrane transport
L-aspartate transmembrane transport
dicarboxylic acid transmembrane transporter activity
organic acid:sodium symporter activity
citrate transmembrane transporter activity
succinate transmembrane transporter activity
L-aspartate transmembrane transporter activity
high-affinity sodium:dicarboxylate symporter activity
sodium:dicarboxylate symporter activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P12330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 499194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 927388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 219684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62300 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792736 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 713907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338261 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540769 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32651 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12330 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P12330 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P12330 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12330 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700930796 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100215 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 54363 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25705 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 26657 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29226 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700930796 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10851805 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851796 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851803 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851808 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851804 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851797 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851802 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851801 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851807 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851800 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851806 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851798 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851799 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10851795 | - | - |
Rat Genome 230 2.0 Array | 1368047_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368047_at | - | - |
Affymetrix HT Rat Focus Array | 1368047_at | - | - |
Affymetrix HT Rat Focus Array | 1368047_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300036698.rn.1 | TC0300004779.rn.1 | RTA TC |
Rat Expression Set 230 | 1368047_PM_at | - | - |
Rat Expression Set 230 | 1368047_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10851795 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21794702 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1350578 | - | - |
RatRef-12 Expression BeadChip | ILMN_56539 | - | - |
