Assay Details
Gene Symbol: Dab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024159.1 | NP_077073.1 | 3 - 4 | 335 | 100 | |
GenBank mRNA | BC097314.1 | - | 3 - 4 | 370 | 7444786 | 100 |
FQ224185.1 | - | 645 | 100 | |||
U95177.1 | - | 3 - 4 | 335 | 100 | ||
U95178.1 | - | 3 - 4 | 335 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis involved in differentiation
in utero embryonic development
positive regulation of receptor recycling
positive regulation of protein phosphorylation
endocytosis
receptor-mediated endocytosis
pinocytosis
apoptotic process
integrin-mediated signaling pathway
activation of JUN kinase activity
endoderm development
excretion
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of neuron projection development
protein transport
Wnt signaling pathway
myeloid cell differentiation
negative regulation of cell growth
positive regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of protein binding
positive regulation of aldosterone biosynthetic process
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of transcription elongation from RNA polymerase II promoter
leading edge cell differentiation
negative regulation of apoptotic process
positive regulation of cell adhesion
positive regulation of endocytosis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
clathrin coat assembly
response to steroid hormone
negative regulation of epithelial cell proliferation
positive regulation of SMAD protein import into nucleus
negative regulation of androgen receptor signaling pathway
negative regulation of ERK1 and ERK2 cascade
cellular response to epidermal growth factor stimulus
cellular response to transforming growth factor beta stimulus
negative regulation of canonical Wnt signaling pathway
positive regulation of substrate adhesion-dependent cell spreading
response to salt
negative regulation of protein localization to plasma membrane
positive regulation of Wnt signaling pathway, planar cell polarity pathway
regulation of Rho-dependent protein serine/threonine kinase activity
positive regulation of clathrin-mediated endocytosis
positive regulation of early endosome to late endosome transport
positive regulation of aldosterone secretion
positive regulation of integrin-mediated signaling pathway
negative regulation of extrinsic apoptotic signaling pathway
integrin binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
protein C-terminus binding
clathrin binding
phosphatidylinositol binding
AP-2 adaptor complex binding
clathrin adaptor activity
cargo receptor activity
SMAD binding
cell morphogenesis involved in differentiation
in utero embryonic development
positive regulation of receptor recycling
positive regulation of protein phosphorylation
endocytosis
receptor-mediated endocytosis
pinocytosis
apoptotic process
integrin-mediated signaling pathway
activation of JUN kinase activity
endoderm development
excretion
positive regulation of epithelial to mesenchymal transition
positive regulation of pathway-restricted SMAD protein phosphorylation
negative regulation of neuron projection development
protein transport
Wnt signaling pathway
myeloid cell differentiation
negative regulation of cell growth
positive regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of protein binding
positive regulation of aldosterone biosynthetic process
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
positive regulation of transcription elongation from RNA polymerase II promoter
leading edge cell differentiation
negative regulation of apoptotic process
positive regulation of cell adhesion
positive regulation of endocytosis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
clathrin coat assembly
response to steroid hormone
negative regulation of epithelial cell proliferation
positive regulation of SMAD protein import into nucleus
negative regulation of androgen receptor signaling pathway
negative regulation of ERK1 and ERK2 cascade
cellular response to epidermal growth factor stimulus
cellular response to transforming growth factor beta stimulus
negative regulation of canonical Wnt signaling pathway
positive regulation of substrate adhesion-dependent cell spreading
response to salt
negative regulation of protein localization to plasma membrane
positive regulation of Wnt signaling pathway, planar cell polarity pathway
regulation of Rho-dependent protein serine/threonine kinase activity
positive regulation of clathrin-mediated endocytosis
positive regulation of early endosome to late endosome transport
positive regulation of aldosterone secretion
positive regulation of integrin-mediated signaling pathway
negative regulation of extrinsic apoptotic signaling pathway
integrin binding
protein binding
phosphatidylinositol-4,5-bisphosphate binding
protein C-terminus binding
clathrin binding
phosphatidylinositol binding
AP-2 adaptor complex binding
clathrin adaptor activity
cargo receptor activity
SMAD binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P513385 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P144944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654209 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 949207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163288 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13877 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144944 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P513385 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P513385 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P144944 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P144944 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P513385 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P513385 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700289744 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701525465 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701826413 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 40015 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100275 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701525465 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700631542 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700289744 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701826413 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10813366 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813365 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813379 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813367 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813369 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813368 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813378 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813362 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813374 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813363 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813364 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813376 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813370 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813377 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813375 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813371 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10813361 | - | - |
Rat Genome 230 2.0 Array | 1368202_a_at | - | - |
Rat Genome 230 2.0 Array | 1368202_PM_a_at | - | - |
Rat Genome 230 2.0 Array | 1372031_at | - | - |
Rat Genome 230 2.0 Array | 1372031_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368202_a_at | - | - |
Affymetrix HT Rat Focus Array | 1368202_PM_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200002980.rn.1 | TC0200000503.rn.1 | RTA TC |
Rat Expression Set 230 | 1368202_a_at | - | - |
Rat Expression Set 230 | 1368202_PM_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813361 | - | - |
Rat Genome U34 Set | U95178_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21977010 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1355383 | - | - |
RatRef-12 Expression BeadChip | ILMN_62287 | - | - |
