Assay Details
| Gene Symbol: Hmox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024387.2 | NP_077363.1 | 1 - 2 | 152 | 107 | |
| GenBank mRNA | J05405.1 | - | 1 - 2 | 144 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the J05405.1, NM_024387.2 transcripts.
Set Membership: |
|

Hmox2
Genomic Map