Assay Details
Gene Symbol: Hsd17b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024392.2 | NP_077368.2 | 17 - 18 | 1552 | 74 | |
GenBank mRNA | BC072472.1 | - | 17 - 18 | 1542 | 7100430 | 74 |
S83279.1 | - | 17 - 18 | 1524 | 74 | ||
U37486.1 | - | 17 - 18 | 1552 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
very long-chain fatty acid metabolic process
osteoblast differentiation
fatty acid beta-oxidation
metabolic process
cholesterol metabolic process
androgen metabolic process
estrogen metabolic process
very long-chain fatty-acyl-CoA metabolic process
medium-chain fatty-acyl-CoA metabolic process
oxidation-reduction process
Sertoli cell development
3-hydroxyacyl-CoA dehydrogenase activity
receptor binding
long-chain-enoyl-CoA hydratase activity
isomerase activity
testosterone dehydrogenase [NAD(P)] activity
3alpha,7alpha,12alpha-trihydroxy-5beta-cholest-24-enoyl-CoA hydratase activity
protein homodimerization activity
17-beta-hydroxysteroid dehydrogenase (NAD+) activity
very long-chain fatty acid metabolic process
osteoblast differentiation
fatty acid beta-oxidation
metabolic process
cholesterol metabolic process
androgen metabolic process
estrogen metabolic process
very long-chain fatty-acyl-CoA metabolic process
medium-chain fatty-acyl-CoA metabolic process
oxidation-reduction process
Sertoli cell development
3-hydroxyacyl-CoA dehydrogenase activity
receptor binding
long-chain-enoyl-CoA hydratase activity
isomerase activity
testosterone dehydrogenase [NAD(P)] activity
3alpha,7alpha,12alpha-trihydroxy-5beta-cholest-24-enoyl-CoA hydratase activity
protein homodimerization activity
17-beta-hydroxysteroid dehydrogenase (NAD+) activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P110009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 252227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690878 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8554 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411910 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847331 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 128847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776086 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31609 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588719 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 526791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548276 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805664 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812303 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170838 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 5132 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P110009 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P110009 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P110009 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P365531 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73842 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 58595 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74402 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94512 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3474 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63184 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6072 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10801663 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801674 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801657 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801658 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801660 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801654 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801664 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801670 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801677 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801672 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801665 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801671 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801661 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801668 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801673 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801666 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801656 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801675 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801676 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801655 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801659 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801667 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801662 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801669 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10801653 | - | - |
Rat Genome 230 2.0 Array | 1367672_PM_at | - | - |
Rat Genome 230 2.0 Array | 1367672_at | - | - |
Affymetrix HT Rat Focus Array | 1367672_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1367672_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800003633.rn.1 | TC1800000492.rn.1 | RTA TC |
Rat Expression Set 230 | 1367672_at | - | - |
Rat Expression Set 230 | 1367672_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10801653 | - | - |
Rat Genome U34 Set | rc_AI013834_s_at | - | - |
Rat Genome U34 Set | S83279_g_at | - | - |
Rat Genome U34 Set | rc_AI101743_s_at | - | - |
Rat Genome U34 Set | S83279_at | - | - |
Rat Genome U34 Set | rc_AI101743_s_at | - | - |
Rat Genome U34 Set | S83279_g_at | - | - |
Rat Genome U34 Set | S83279_at | - | - |
Rat Genome U34 Set | rc_AI013834_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22129348 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360823 | - | - |
RatRef-12 Expression BeadChip | ILMN_55709 | - | - |
