Assay Details
| Gene Symbol: Lss | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031049.1 | NP_112311.1 | 5 - 6 | 75 | ||
| GenBank mRNA | D45252.1 | - | 5 - 6 | 623 | 75 | |
| U31352.1 | - | 5 - 6 | 75 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P126508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235430 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 113144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390707 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539670 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679967 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137182 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582576 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175740 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380091 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285918 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174514 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267921 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85812 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P126508 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P126508 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P126508 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P334218 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700146984 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701744110 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701228305 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92139 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 48683 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 74910 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701744110 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700146984 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701228305 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10832393 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832395 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832384 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832385 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832396 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832403 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832390 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832402 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832398 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832401 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832388 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832399 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832394 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832391 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832382 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832381 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832380 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832387 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832386 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832392 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832389 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832383 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832400 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832379 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832397 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10832378 | - | - |
| Rat Genome 230 2.0 Array | 1372973_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368086_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1372973_at | - | - |
| Rat Genome 230 2.0 Array | 1368086_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1368086_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1368086_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1372973_at | - | - |
| Affymetrix HT Rat Focus Array | 1372973_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR2000010969.rn.1 | TC2000001328.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368086_a_at | - | - |
| Rat Expression Set 230 | 1368086_PM_a_at | - | - |
| Rat Expression Set 230 | 1372973_at | - | - |
| Rat Expression Set 230 | 1372973_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10832378 | - | - |
| Rat Genome U34 Set | D45252_s_at | - | - |
| Rat Genome U34 Set | U31352_at | - | - |
| Rat Genome U34 Set | E12275cds_s_at | - | - |
| Rat Genome U34 Set | rc_AI137259_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21014299 | - | - |

Lss
Genomic Map