Assay Details
Gene Symbol: Pth2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031089.2 | NP_112351.1 | 5 - 6 | 719 | 81 | |
GenBank mRNA | U55836.1 | - | 5 - 6 | 719 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P000201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333235 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103253 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 49039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 87284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 646685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 577997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141984 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 98032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 176940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 36789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81072 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000201 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P000201 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P000201 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12509 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66615 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 65013 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73279 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16199 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29653 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 33721 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14003 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62000 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76632 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10924004 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924007 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924008 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924003 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924009 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924011 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924006 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924010 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924001 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923999 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924000 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924002 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10924005 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10923998 | - | - |
Rat Genome 230 2.0 Array | 1369461_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369461_at | - | - |
Affymetrix HT Rat Focus Array | 1369461_at | - | - |
Affymetrix HT Rat Focus Array | 1369461_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0900006649.rn.1 | TC0900000809.rn.1 | RTA TC |
Rat Expression Set 230 | 1369461_at | - | - |
Rat Expression Set 230 | 1369461_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10923998 | - | - |
Rat Genome U34 Set | U55836_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21963547 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1349499 | - | - |
RatRef-12 Expression BeadChip | ILMN_52227 | - | - |
