Assay Details
Gene Symbol: Prl3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031316.1 | NP_112606.1 | 1 - 2 | 80 | 65 | |
GenBank mRNA | AB019119.1 | - | 1 - 2 | 80 | 65 | |
AB019945.1 | - | 1 - 2 | 71 | 65 | ||
AF150741.1 | - | 1 - 2 | 80 | 65 | ||
AF234638.1 | - | 1 - 2 | 71 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB019119.1, AB019945.1, AF150741.1, AF234638.1, NM_031316.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
