Assay Details
| Gene Symbol: Met | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031517.2 | NP_113705.1 | 10 - 11 | 2586 | 85 | |
| XM_006236129.2 | XP_006236191.1 | 12 - 13 | 3002 | 85 | ||
| GenBank mRNA | U65007.1 | - | 10 - 11 | 2586 | 85 | |
| X96786.1 | - | 10 - 11 | 2586 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Met
Genomic Map