Assay Details
| Gene Symbol: Por | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031576.1 | NP_113764.1 | 220 | 63 | ||
| GenBank mRNA | M10068.1 | - | 1 - 2 | 184 | 63 | |
| M12516.1 | - | 220 | 63 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of growth plate cartilage chondrocyte proliferation
response to nutrient
carnitine metabolic process
flavonoid metabolic process
internal peptidyl-lysine acetylation
fatty acid oxidation
positive regulation of chondrocyte differentiation
positive regulation of monooxygenase activity
response to drug
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
nitrate catabolic process
positive regulation of cholesterol biosynthetic process
positive regulation of smoothened signaling pathway
nitric oxide catabolic process
oxidation-reduction process
negative regulation of lipase activity
demethylation
cellular response to gonadotropin stimulus
cellular response to follicle-stimulating hormone stimulus
cellular response to peptide hormone stimulus
positive regulation of steroid hormone biosynthetic process
regulation of cholesterol metabolic process
cellular organofluorine metabolic process
NADPH-hemoprotein reductase activity
cytochrome-b5 reductase activity, acting on NAD(P)H
iron ion binding
nitric oxide dioxygenase activity
electron carrier activity
FMN binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
hydrolase activity
enzyme binding
iron-cytochrome-c reductase activity
flavin adenine dinucleotide binding
NADP binding
regulation of growth plate cartilage chondrocyte proliferation
response to nutrient
carnitine metabolic process
flavonoid metabolic process
internal peptidyl-lysine acetylation
fatty acid oxidation
positive regulation of chondrocyte differentiation
positive regulation of monooxygenase activity
response to drug
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
nitrate catabolic process
positive regulation of cholesterol biosynthetic process
positive regulation of smoothened signaling pathway
nitric oxide catabolic process
oxidation-reduction process
negative regulation of lipase activity
demethylation
cellular response to gonadotropin stimulus
cellular response to follicle-stimulating hormone stimulus
cellular response to peptide hormone stimulus
positive regulation of steroid hormone biosynthetic process
regulation of cholesterol metabolic process
cellular organofluorine metabolic process
NADPH-hemoprotein reductase activity
cytochrome-b5 reductase activity, acting on NAD(P)H
iron ion binding
nitric oxide dioxygenase activity
electron carrier activity
FMN binding
oxidoreductase activity
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
hydrolase activity
enzyme binding
iron-cytochrome-c reductase activity
flavin adenine dinucleotide binding
NADP binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P402375 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 165170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936936 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536961 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 703059 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823100 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823871 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153150 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502692 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 829155 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207054 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472503 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 523266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 182156 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699812 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317602 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351501 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 528816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64998 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50990 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180778 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62473 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P402375 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P402375 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P402375 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P402375 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100549 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 95988 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66738 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69102 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 89513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10761170 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761169 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761173 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761163 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761179 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761176 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761175 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761166 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761164 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761178 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761172 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761171 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761174 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761177 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761168 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761167 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10761162 | - | - |
| Rat Genome 230 2.0 Array | 1387109_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368213_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387109_at | - | - |
| Rat Genome 230 2.0 Array | 1368213_at | - | - |
| Affymetrix HT Rat Focus Array | 1387109_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1368213_at | - | - |
| Affymetrix HT Rat Focus Array | 1387109_at | - | - |
| Affymetrix HT Rat Focus Array | 1368213_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1200011590.rn.1 | TC1200001346.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200011594.rn.1 | TC1200001346.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR1200011592.rn.1 | TC1200001346.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387109_PM_at | - | - |
| Rat Expression Set 230 | 1368213_at | - | - |
| Rat Expression Set 230 | 1368213_PM_at | - | - |
| Rat Expression Set 230 | 1387109_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10761162 | - | - |
| Rat Genome U34 Set | M10068mRNA_s_at | - | - |
| Rat Genome U34 Set | E01524cds_s_at | - | - |
| Rat Genome U34 Set | rc_AI137856_s_at | - | - |
| Rat Genome U34 Set | rc_AA956507_at | - | - |
| Rat Genome U34 Set | rc_AI137856_s_at | - | - |
| Rat Genome U34 Set | rc_AA956507_at | - | - |
| Rat Genome U34 Set | M10068mRNA_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21758989 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358080 | - | - |
| RatRef-12 Expression BeadChip | ILMN_66196 | - | - |

Por
Genomic Map