Assay Details
Gene Symbol: Grb14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031623.1 | NP_113811.1 | 8 - 9 | 1088 | 77 | |
GenBank mRNA | AF076619.1 | - | 8 - 9 | 1088 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P020627 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P074918 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75883 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 915585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 469940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 434964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 180424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 442796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619520 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 908436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 234637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266315 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 554882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541397 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799017 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 962400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 776254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696569 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 335684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 109179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61818 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074918 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P020627 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P020627 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P074918 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P020627 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P074918 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P12617 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701918105 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92270 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30274 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71806 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71859 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45130 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44547 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100990 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62660 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701918105 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10845761 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845756 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845757 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845754 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845758 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845752 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845763 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845753 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845759 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845764 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845755 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845762 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845760 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10845751 | - | - |
Rat Genome 230 2.0 Array | 1368618_at | - | - |
Rat Genome 230 2.0 Array | 1368618_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368618_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1368618_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300024897.rn.1 | TC0300003206.rn.1 | RTA TC |
Rat Expression Set 230 | 1368618_PM_at | - | - |
Rat Expression Set 230 | 1368618_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10845751 | - | - |
Rat Genome U34 Set | AF076619_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21012749 | - | - |
