Assay Details
| Gene Symbol: Casp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031632.1 | NP_113820.1 | 1 - 2 | 180 | 68 | |
| GenBank mRNA | AF262319.1 | - | 1 - 2 | 180 | 68 | |
| AF271996.1 | - | 1 - 2 | 147 | 68 | ||
| AF286006.1 | - | 1 - 2 | 180 | 68 | ||
| AF293333.1 | - | 1 - 2 | 180 | 68 | ||
| AF308469.1 | - | 1 - 2 | 147 | 68 | ||
| AF517560.1 | - | 1 - 2 | 180 | 68 | ||
| AY008275.1 | - | 1 - 2 | 147 | 68 | ||
| AY027667.1 | - | 1 - 2 | 156 | 68 | ||
| AY124461.1 | - | 1 - 2 | 180 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp9
Genomic Map