Assay Details
Gene Symbol: B4galt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031740.1 | NP_113928.1 | 2 | 954 | 84 | |
GenBank mRNA | AF048687.1 | - | 2 | 954 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P502287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 543579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 383927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387708 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 21968 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 179565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157568 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P502287 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P502287 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P502287 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1032732 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P502287 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700818766 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53509 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 66788 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43265 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68444 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 700818766 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10803399 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803395 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803402 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803404 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803396 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803398 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803400 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803403 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803401 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803397 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10803394 | - | - |
Rat Genome 230 2.0 Array | 1387206_at | - | - |
Rat Genome 230 2.0 Array | 1387206_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387206_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1387206_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800000960.rn.1 | TC1800000137.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800000961.rn.1 | TC1800000137.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800000963.rn.1 | TC1800000137.rn.1 | RTA TC |
Rat Expression Set 230 | 1387206_PM_at | - | - |
Rat Expression Set 230 | 1387206_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10803394 | - | - |
Rat Genome U34 Set | AF048687_s_at | - | - |
Rat Genome U34 Set | rc_AI059686_s_at | - | - |
Rat Genome U34 Set | rc_AA964073_s_at | - | - |
Rat Genome U34 Set | AF048687_s_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22410634 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376522 | - | - |
