Assay Details
| Gene Symbol: Gda | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031776.2 | NP_113964.2 | 10 - 11 | 1035 | 78 | |
| GenBank mRNA | AF026472.2 | - | 10 - 11 | 1092 | 78 | |
| AF245172.1 | - | 10 - 11 | 1035 | 78 | ||
| FQ233257.1 | - | 10 - 11 | 1058 | 78 | ||
| FQ233523.1 | - | 10 - 11 | 1063 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P129048 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608240 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 548106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691825 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199052 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210105 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71907 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593916 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405827 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 185540 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 930330 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 784004 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265062 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362453 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584553 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562677 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763510 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858281 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305758 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689469 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 263311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763810 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495214 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163472 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773446 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406904 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599483 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286814 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492089 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96770 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 1110 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9265 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148931 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73989 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91595 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P129048 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P129048 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P129048 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P402750 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 702150070 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92435 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 73832 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 96370 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 55094 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 50074 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 30609 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 702150070 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10729319 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729328 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729322 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729325 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729329 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729316 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729327 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729324 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729317 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729320 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729330 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729321 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729323 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729326 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729318 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10729314 | - | - |
| Rat Genome 230 2.0 Array | 1372011_at | - | - |
| Rat Genome 230 2.0 Array | 1387659_at | - | - |
| Rat Genome 230 2.0 Array | 1372011_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387659_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387659_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387659_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0100059781.rn.1 | TC0100007398.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387659_PM_at | - | - |
| Rat Expression Set 230 | 1387659_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10729314 | - | - |
| Rat Genome U34 Set | U03629_at | - | - |
| Rat Genome U34 Set | rc_AA859837_g_at | - | - |
| Rat Genome U34 Set | rc_AA859837_at | - | - |
| Rat Genome U34 Set | rc_AA964867_at | - | - |
| Rat Genome U34 Set | rc_AI236955_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21194218 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357452 | - | - |
| RatRef-12 Expression BeadChip | ILMN_65816 | - | - |

Gda
Genomic Map