Assay Details
| Gene Symbol: Agxt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031835.2 | NP_114023.1 | 3 - 4 | 394 | 74 | |
| XM_006232069.2 | XP_006232131.1 | 3 - 4 | 462 | 74 | ||
| GenBank mRNA | AB002584.1 | - | 3 - 4 | 74 | ||
| BC081765.1 | - | 3 - 4 | 394 | 7106995 | 74 | |
| D38100.1 | - | 3 - 4 | 74 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
glyoxylate catabolic process
glycine biosynthetic process, by transamination of glyoxylate
L-alanine catabolic process, by transamination
positive regulation of nitric oxide biosynthetic process
alanine-glyoxylate transaminase activity
beta-alanine-pyruvate transaminase activity
pyridoxal phosphate binding
(R)-3-amino-2-methylpropionate-pyruvate transaminase activity
glyoxylate catabolic process
glycine biosynthetic process, by transamination of glyoxylate
L-alanine catabolic process, by transamination
positive regulation of nitric oxide biosynthetic process
alanine-glyoxylate transaminase activity
beta-alanine-pyruvate transaminase activity
pyridoxal phosphate binding
(R)-3-amino-2-methylpropionate-pyruvate transaminase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P313644 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230925 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 644581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735196 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253433 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 709973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 584632 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300297 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778930 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6206 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912772 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10876 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19737 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472465 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539895 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731227 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736843 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 194579 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172988 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779524 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217131 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218437 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94559 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49554 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P313644 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P313644 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P313644 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P313644 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 88123 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 91427 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 6722 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10813653 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813647 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813661 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813663 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813651 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813650 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813657 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813648 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813652 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813654 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813649 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813658 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813660 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813656 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813662 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813659 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10813646 | - | - |
| Rat Genome 230 2.0 Array | 1368659_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368659_at | - | - |
| Affymetrix HT Rat Focus Array | 1368659_at | - | - |
| Affymetrix HT Rat Focus Array | 1368659_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200003517.rn.1 | TC0200000560.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368659_at | - | - |
| Rat Expression Set 230 | 1368659_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10813646 | - | - |
| Rat Genome U34 Set | AB002584_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21431557 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1362683 | - | - |
| RatRef-12 Expression BeadChip | ILMN_58445 | - | - |

Agxt2
Genomic Map