Assay Details
| Gene Symbol: Sept9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113497.1 | NP_001106969.1 | 255 | 70 | ||
| NM_031837.2 | NP_114025.2 | 1143 | 70 | |||
| NM_176856.2 | NP_789826.2 | 1262 | 70 | |||
| GenBank mRNA | AF170253.1 | - | 1263 | 70 | ||
| AF173899.1 | - | 1142 | 70 | |||
| AF180525.1 | - | 2 - 3 | 422 | 70 | ||
| AF180526.1 | - | 243 | 70 | |||
| BC166602.1 | - | 808 | 70 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P393282 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P085405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879086 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826729 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772120 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567026 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533894 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760058 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 583266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283010 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314389 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482180 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 145286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593690 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849882 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395001 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108581 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786334 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59575 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98401 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891712 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 616445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608645 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559905 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290060 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761955 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667093 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101402 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 374964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357385 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810672 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859296 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425273 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289899 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638942 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643563 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 632987 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868476 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 439013 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787162 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289601 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641964 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233053 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 502079 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450983 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685421 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 81210 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151499 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671021 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 352656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104333 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328373 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811972 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34383 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 315418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769742 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46357 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 658381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362759 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357572 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29652 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67747 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163066 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170841 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84289 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 24239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126095 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170926 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6161 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085405 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P393282 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P393282 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P085405 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P393282 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P085405 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P393282 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700628320 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68879 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103725 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 69927 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 24091 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76117 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40918 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51531 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 92530 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 93268 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700628320 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10739894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739890 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739888 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739896 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739893 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739899 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739891 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739889 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739901 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739902 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739897 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739898 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739900 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739895 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739892 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739903 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10739886 | - | - |
| Rat Genome 230 2.0 Array | 1367698_PM_a_at | - | - |
| Rat Genome 230 2.0 Array | 1367698_a_at | - | - |
| Rat Genome 230 2.0 Array | 1386906_a_at | - | - |
| Rat Genome 230 2.0 Array | 1386906_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1386906_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1367698_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1367698_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1386906_PM_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1000018016.rn.1 | TC1000001995.rn.1 | RTA TC |
| Rat Expression Set 230 | 1386906_a_at | - | - |
| Rat Expression Set 230 | 1367698_a_at | - | - |
| Rat Expression Set 230 | 1386906_PM_a_at | - | - |
| Rat Expression Set 230 | 1367698_PM_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10739886 | - | - |
| Rat Genome U34 Set | rc_AA819530_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22352725 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376484 | - | - |
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1375516 | - | - |
| RatRef-12 Expression BeadChip | ILMN_68343 | - | - |
| RatRef-12 Expression BeadChip | ILMN_52171 | - | - |

Sept9
Genomic Map