Assay Details
Gene Symbol: Ptprv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033099.1 | NP_149090.1 | 19 - 20 | 3579 | 77 | |
XM_008769570.1 | XP_008767792.1 | 19 - 20 | 3540 | 77 | ||
GenBank mRNA | L36884.1 | - | 19 - 20 | 3579 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P063681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207242 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291673 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27633 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773290 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114124 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59030 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26617 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P063681 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P063681 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P063681 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P787160 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14771 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 80010 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10767894 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767879 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767890 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767891 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767899 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767892 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767911 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767900 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767909 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767887 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767880 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767906 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767905 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767893 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767877 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767888 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767895 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767897 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767907 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767896 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767886 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767901 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767903 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767881 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767882 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767878 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767884 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767885 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767898 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767904 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767889 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767908 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767902 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10764192 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10767876 | - | - |
Rat Genome 230 2.0 Array | 1387136_PM_at | - | - |
Rat Genome 230 2.0 Array | 1387136_at | - | - |
Affymetrix HT Rat Focus Array | 1387136_at | - | - |
Affymetrix HT Rat Focus Array | 1387136_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1300009879.rn.1 | TC1300001455.rn.1 | RTA TC |
Rat Expression Set 230 | 1387136_PM_at | - | - |
Rat Expression Set 230 | 1387136_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10767876 | - | - |
Rat Genome U34 Set | L36884_at | - | - |
Rat Genome U34 Set | L36884_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21488127 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369422 | - | - |
RatRef-12 Expression BeadChip | ILMN_63614 | - | - |
