Assay Details
| Gene Symbol: Kcnk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033376.1 | NP_203694.1 | 1 - 2 | 383 | 87 | |
| GenBank mRNA | AB048823.1 | - | 2 - 3 | 236 | 87 | |
| AB048824.1 | - | 2 - 3 | 225 | 87 | ||
| AF031384.1 | - | 1 - 2 | 383 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB048824.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
brain development
ion transmembrane transport
regulation of ion transmembrane transport
response to drug
negative regulation of cytosolic calcium ion concentration
cellular response to zinc ion
cellular response to hypoxia
potassium ion transmembrane transport
cochlea development
ion channel activity
open rectifier potassium channel activity
protein binding
protein C-terminus binding
potassium ion leak channel activity
protein homodimerization activity
S100 protein binding
protein heterodimerization activity
brain development
ion transmembrane transport
regulation of ion transmembrane transport
response to drug
negative regulation of cytosolic calcium ion concentration
cellular response to zinc ion
cellular response to hypoxia
potassium ion transmembrane transport
cochlea development
ion channel activity
open rectifier potassium channel activity
protein binding
protein C-terminus binding
potassium ion leak channel activity
protein homodimerization activity
S100 protein binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P036900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617858 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818157 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58722 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278909 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556234 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366181 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672466 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433286 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973475 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131141 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786117 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73991 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691070 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141818 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115188 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27660 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036900 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P036900 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P036900 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P671477 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700183736 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 37797 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90749 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 46250 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700183736 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10889041 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889038 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889037 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889040 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889039 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10889036 | - | - |
| Rat Genome 230 2.0 Array | 1375807_at | - | - |
| Rat Genome 230 2.0 Array | 1375807_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387441_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389610_at | - | - |
| Rat Genome 230 2.0 Array | 1398313_a_at | - | - |
| Rat Genome 230 2.0 Array | 1387441_at | - | - |
| Rat Genome 230 2.0 Array | 1389610_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1398313_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1387441_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387441_at | - | - |
| Affymetrix HT Rat Focus Array | 1398313_PM_a_at | - | - |
| Affymetrix HT Rat Focus Array | 1375807_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1375807_at | - | - |
| Affymetrix HT Rat Focus Array | 1398313_a_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0600014925.rn.1 | TC0600002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600014928.rn.1 | TC0600002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600014924.rn.1 | TC0600002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600014923.rn.1 | TC0600002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600014926.rn.1 | TC0600002034.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0600014927.rn.1 | TC0600002034.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387441_at | - | - |
| Rat Expression Set 230 | 1375807_PM_at | - | - |
| Rat Expression Set 230 | 1375807_at | - | - |
| Rat Expression Set 230 | 1398313_PM_a_at | - | - |
| Rat Expression Set 230 | 1387441_PM_at | - | - |
| Rat Expression Set 230 | 1398313_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10889036 | - | - |
| Rat Genome U34 Set | rc_AA925138_s_at | - | - |
| Rat Genome U34 Set | AF031384_at | - | - |
| Rat Genome U34 Set | AF031384_at | - | - |
| Rat Genome U34 Set | rc_AA925138_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22153441 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1364257 | - | - |
| RatRef-12 Expression BeadChip | ILMN_66193 | - | - |

Kcnk3
Genomic Map