Assay Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053416.2 | NP_445868.2 | 9 - 10 | 1215 | 94 | |
| XM_006234108.2 | XP_006234170.2 | 10 - 11 | 1553 | 94 | ||
| XM_008761777.1 | XP_008759999.1 | 10 - 11 | 1253 | 94 | ||
| XM_008761778.1 | XP_008760000.1 | 10 - 11 | 1253 | 94 | ||
| GenBank mRNA | BC078774.1 | - | 8 - 9 | 837 | 94 | |
| DY547419.1 | - | 1 - 2 | 83 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P158484 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 296617 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 564057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 441198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192319 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348753 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690534 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716923 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687244 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 375366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896498 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824183 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12329 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895259 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297593 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596144 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594606 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610860 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465366 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196224 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 943889 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 879968 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 601231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80943 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414611 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76000 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14668 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251022 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812432 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732789 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27138 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 328646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 91727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707852 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13047 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569035 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529247 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 289098 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397622 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 805787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529168 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54124 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203165 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30557 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756664 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 221507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137390 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 489640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7880 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847801 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269509 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 82689 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 179616 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 88813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71751 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27504 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47372 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173792 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159064 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 22133 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123690 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158484 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P158484 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P158484 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P245636 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 508 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 78632 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 77882 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34749 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57564 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 62176 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8920 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19320 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10844819 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844817 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844810 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844812 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844816 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844824 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844820 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844809 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844814 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844811 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844813 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844821 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844822 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844818 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844815 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844823 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10844808 | - | - |
| Rat Genome 230 2.0 Array | 1380763_at | - | - |
| Rat Genome 230 2.0 Array | 1380763_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368903_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1368903_at | - | - |
| Affymetrix HT Rat Focus Array | 1368903_at | - | - |
| Affymetrix HT Rat Focus Array | 1368903_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300023228.rn.1 | TC0300002994.rn.1 | RTA TC |
| Rat Expression Set 230 | 1368903_PM_at | - | - |
| Rat Expression Set 230 | 1368903_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10844808 | - | - |
| Rat Genome U34 Set | rc_AA858834_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21651232 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365116 | - | - |
| RatRef-12 Expression BeadChip | ILMN_63605 | - | - |

Strbp
Genomic Map