Assay Details
| Gene Symbol: Slc7a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053442.1 | NP_445894.1 | 7 - 8 | 1622 | 78 | |
| GenBank mRNA | AB024400.1 | - | 7 - 8 | 1622 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
amino acid transport
organic cation transport
neutral amino acid transport
L-amino acid transport
toxin transport
L-alpha-amino acid transmembrane transport
organic cation transmembrane transporter activity
amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
toxin transporter activity
peptide antigen binding
amino acid transport
organic cation transport
neutral amino acid transport
L-amino acid transport
toxin transport
L-alpha-amino acid transmembrane transport
organic cation transmembrane transporter activity
amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
toxin transporter activity
peptide antigen binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P124050 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89252 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130993 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446959 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724875 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828228 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 571568 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961427 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 142127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123134 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781269 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712313 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131655 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911511 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875584 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250523 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322615 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749807 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84032 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222608 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141046 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382715 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951647 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 535239 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744477 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248367 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617208 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744328 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243781 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235169 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578745 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 701950 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426497 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756859 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539346 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 719494 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149217 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952853 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479746 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69929 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54044 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32714 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17015 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131818 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P124050 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P124050 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P124050 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P606971 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 19589 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 26118 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 97004 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 103277 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 60977 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10783649 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783655 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783659 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783651 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783656 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783654 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783658 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783657 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783652 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783650 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783653 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10783648 | - | - |
| Rat Genome 230 2.0 Array | 1387057_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387057_at | - | - |
| Affymetrix HT Rat Focus Array | 1387057_at | - | - |
| Affymetrix HT Rat Focus Array | 1387057_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1500010728.rn.1 | TC1500001679.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387057_PM_at | - | - |
| Rat Expression Set 230 | 1368110_a_at | - | - |
| Rat Expression Set 230 | 1387057_at | - | - |
| Rat Expression Set 230 | 1368110_PM_a_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10783648 | - | - |
| Rat Genome U34 Set | rc_AA899235_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22185377 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1373072 | - | - |
| RatRef-12 Expression BeadChip | ILMN_57185 | - | - |

Slc7a8
Genomic Map