Assay Details
Gene Symbol: Ppp1r9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053473.2 | NP_445925.1 | 7 - 8 | 2324 | 99 | |
XM_006236057.2 | XP_006236119.1 | 9 - 10 | 2501 | 99 | ||
XM_006236058.2 | XP_006236120.1 | 9 - 10 | 2541 | 99 | ||
XM_006236061.2 | XP_006236123.1 | 9 - 10 | 2501 | 99 | ||
XM_006236063.2 | XP_006236125.1 | 8 - 9 | 2435 | 99 | ||
XM_006236064.2 | XP_006236126.1 | 8 - 9 | 2435 | 99 | ||
XM_006236066.2 | XP_006236128.1 | 9 - 10 | 2498 | 99 | ||
XM_006236068.2 | XP_006236130.1 | 8 - 9 | 2431 | 99 | ||
GenBank mRNA | U72994.1 | - | 7 - 8 | 2328 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P451638 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 401449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175471 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 223772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917341 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698637 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552394 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43768 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811536 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 409137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 453233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941344 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 604221 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34808 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850782 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95889 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 188579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 140429 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518943 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28712 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799281 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310318 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 614938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655542 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372940 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838781 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 537592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 359816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 488831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240786 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 260025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587885 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862343 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376746 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 172871 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 137127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834139 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641347 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 637002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351415 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 242159 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 700950 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69818 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582606 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673358 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346501 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 692076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919324 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 190346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 368527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34847 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17765 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 171168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168251 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 121495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 26935 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79329 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P451638 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P451638 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P451638 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P451638 | - | - |
Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701531085 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1900 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102992 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89938 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49366 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64251 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6532 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38919 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62433 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 5157 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42562 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36761 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14836 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84615 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29277 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102139 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8871 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67358 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29940 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9136 | - | - |
Agilent Rat cDNA Microarray (G4105A) | 701531085 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10853620 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853625 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853623 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853632 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853618 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853626 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853616 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853617 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853624 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853631 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853615 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853627 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853633 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853622 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853629 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853619 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10853614 | - | - |
Rat Genome 230 2.0 Array | 1369095_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369095_at | - | - |
Affymetrix HT Rat Focus Array | 1369095_at | - | - |
Affymetrix HT Rat Focus Array | 1369095_PM_at | - | - |
Rat Expression Set 230 | 1369095_at | - | - |
Rat Expression Set 230 | 1369095_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10853614 | - | - |
Rat Genome U34 Set | rc_AI145444_at | - | - |
Rat Genome U34 Set | rc_AI112825_at | - | - |
Rat Genome U34 Set | rc_AI145444_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21155262 | - | - |
